APO455831837 Apollo Scientific Ltd

5-Methylthiophene-2-boronic acid

  • MDL number MFCD01318166
  • Cas Number : 162607-20-7
  • Molecular Weight : 141.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 160-165(dec.)
Iupac Name 5-methyl-2-thienylboronic acid
Inchi Code 1S/C5H7BO2S/c1-4-2-3-5(9-4)6(7)8/h2-3,7-8H,1H3
Inchi Key NRIYPIBRPGAWDD-UHFFFAOYSA-N

Description

Synonym 2-Borono-5-methylthiophene
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 160-165(dec.)
Product Link https://store.apolloscientific.co.uk/product/5-methylthiophene-2-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylthiophene-2-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart