APO455831838 Apollo Scientific Ltd

1-(Pyridin-2-yl)-N-[(pyridin-3-yl)methyl]methylamine

  • MDL number MFCD02656617
  • Cas Number : 128802-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3'-(Iminodimethanediyl)dipyridine
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31011

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31011
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart