APO455831846 Apollo Scientific Ltd

4-Bromo-N-methylbenzenesulphonamide

  • MDL number MFCD00993162
  • Cas Number : 703-12-8
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 70-72
Iupac Name 4-bromo-N-methylbenzenesulfonamide
Inchi Code 1S/C7H8BrNO2S/c1-9-12(10,11)7-4-2-6(8)3-5-7/h2-5,9H,1H3
Inchi Key ZAHMEHGOFNLRQN-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 70-72
Product Link https://store.apolloscientific.co.uk/product/4-bromo-n-methylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-n-methylbenzenesulphonamide
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart