APO455831848 Apollo Scientific Ltd

4-Bromo-N-(2-hydroxyethyl)benzenesulphonamide

  • MDL number MFCD03470703
  • Cas Number : 59724-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-78
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31023

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31023
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart