APO455831851 Apollo Scientific Ltd

4-Benzyloxyindole-2-boronic acid, N-BOC protected

  • MDL number MFCD04973739
  • Cas Number : 850568-52-4
  • Molecular Weight : 367.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 102-104
Iupac Name 4-(benzyloxy)-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid
Inchi Code 1S/C20H22BNO5/c1-20(2,3)27-19(23)22-16-10-7-11-17(15(16)12-18(22)21(24)25)26-13-14-8-5-4-6-9-14/h4-12,24-25H,13H2,1-3H3
Inchi Key YXJBIADEQVCONT-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 102-104
Product Link https://store.apolloscientific.co.uk/product/4-benzyloxyindole-2-boronic-acid-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/4-benzyloxyindole-2-boronic-acid-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart