APO455831862 Apollo Scientific Ltd

5-Amino-2-bromopyridine, 5-BOC protected

  • MDL number MFCD07363808
  • Cas Number : 218594-15-1
  • Molecular Weight : 273.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-126
Synonym tert-Butyl (6-bromopyridin-3-yl)carbamate
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-bromopyridine-5-boc-protected

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-bromopyridine-5-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart