APO455831867 Apollo Scientific Ltd

1-Methylpiperidine-3-carboxylic acid

  • MDL number MFCD08059558
  • Cas Number : 5657-70-5
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 162-163
Iupac Name 1-methyl-3-piperidinecarboxylic acid
Inchi Code 1S/C7H13NO2/c1-8-4-2-3-6(5-8)7(9)10/h6H,2-5H2,1H3,(H,9,10)
Inchi Key AYIXGVABNMIOLK-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-1-methylpiperidine, 1-Methylnipecotic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 162-163
Product Link https://store.apolloscientific.co.uk/product/1-methylpiperidine-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylpiperidine-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart