APO455831868 Apollo Scientific Ltd

3-(Naphth-2-yl)-1H-pyrazole

  • MDL number MFCD01569444
  • Cas Number : 150433-20-8
  • Molecular Weight : 194.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-159
Synonym 2-(1H-Pyrazol-3-yl)naphthalene
Density 1.302
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-naphth-2-yl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-naphth-2-yl-1h-pyrazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart