APO455831870 Apollo Scientific Ltd

1,3'-Carbonyldi(piperidine)

  • MDL number MFCD01716373
  • Cas Number : 40576-21-4
  • Molecular Weight : 196.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-piperidinylcarbonyl)piperidine
Inchi Code 1S/C11H20N2O/c14-11(10-5-4-6-12-9-10)13-7-2-1-3-8-13/h10,12H,1-9H2
Inchi Key OSZRYTYCKGZYLB-UHFFFAOYSA-N

Description

Synonym Piperidin-1-yl(piperidin-3-yl)methanone
Flash Point 120-125/0.3mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/13-carbonyldipiperidine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-carbonyldipiperidine
pictograms GHS07
hstatements H302;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart