APO455831877 Apollo Scientific Ltd

1-Benzyl-5-methyl-1H-1,2,3-triazole-4-carbonyl chloride

  • MDL number MFCD07780129
  • Cas Number : 664351-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Synonym 1-Benzyl-4-(chloroformyl)-5-methyl-1H-1,2,3-triazole, 5-Methyl-1-(phenylmethyl)-1H-1,2,3-triazole-4-carbonyl chloride
Purity 97
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31099

Safety Information

signalword Danger
pcodes P280;P260;P308+P313
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31099
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart