APO455831892 Apollo Scientific Ltd

5-Acetamido-2-bromopyridine

  • MDL number MFCD07363807
  • Cas Number : 29958-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-140
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31085

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31085
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart