APO455831895 Apollo Scientific Ltd

2-Amino-6-chloro-3-nitropyridine

  • MDL number MFCD00044350
  • Cas Number : 27048-04-0
  • Molecular Weight : 173.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 194-196
Iupac Name 6-chloro-3-nitro-2-pyridinylamine
Inchi Code 1S/C5H4ClN3O2/c6-4-2-1-3(9(10)11)5(7)8-4/h1-2H,(H2,7,8)
Inchi Key WERABQRUGJIMKQ-UHFFFAOYSA-N

Description

Synonym 6-Chloro-3-nitropyridin-2-amine
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 194-196
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-chloro-3-nitropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-chloro-3-nitropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart