APO455831901 Apollo Scientific Ltd

1,2-Dihydro-4-methoxy-2-oxopyridine-3-carbonitrile

  • MDL number MFCD00975442
  • Cas Number : 21642-98-8
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 275-276
Iupac Name 2-hydroxy-4-methoxynicotinonitrile
Inchi Code 1S/C7H6N2O2/c1-11-6-2-3-9-7(10)5(6)4-8/h2-3H,1H3,(H,9,10)
Inchi Key MWGIDWPSRDMIQN-UHFFFAOYSA-N

Description

Synonym 3-Cyano-1,2-dihydro-4-methoxy-2-oxopyridine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 275-276
Product Link https://store.apolloscientific.co.uk/product/12-dihydro-4-methoxy-2-oxopyridine-3-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/12-dihydro-4-methoxy-2-oxopyridine-3-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart