APO455831909 Apollo Scientific Ltd

5-Amino-3H-1,2,4-dithiazole-3-thione

  • MDL number MFCD00051660
  • Cas Number : 6846-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-203
Synonym 3-Amino-3H-1,2,4-dithiazole-5-thione, Xanthane Hydride
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12640

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12640
pictograms GHS07;GHS08
hstatements H302;H315;H319;H335;H373
Availability details for
Quantity
  • Availability
  • Add to Cart