APO455831910 Apollo Scientific Ltd

3-Bromo-2,6-dichloropyridine

  • MDL number MFCD06798231
  • Cas Number : 866755-20-6
  • Molecular Weight : 226.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 68-71
Iupac Name 3-bromo-2,6-dichloropyridine
Inchi Code 1S/C5H2BrCl2N/c6-3-1-2-4(7)9-5(3)8/h1-2H
Inchi Key PZHASTRIYXMWKM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 68-71
Product Link https://store.apolloscientific.co.uk/product/3-bromo-26-dichloropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-26-dichloropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart