APO455831911 Apollo Scientific Ltd

N,N-Diethyl-4-methylaniline

  • MDL number MFCD00015113
  • Cas Number : 613-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Diethyl-p-toluidine, 4-(Diethylamino)toluene
Density 0.92
Flash Point 102
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 230
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16075

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16075
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart