APO455831913 Apollo Scientific Ltd

3-Acetyl-6-bromocoumarin

  • MDL number MFCD00024075
  • Cas Number : 2199-93-1
  • Molecular Weight : 267.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-229
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-acetyl-6-bromocoumarin

Safety Information

signalword Warning
pcodes P280;P301+P312;P261
MSDS https://store.apolloscientific.co.uk/product/3-acetyl-6-bromocoumarin
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart