APO455831927 Apollo Scientific Ltd

1,2-Dihydro-4-oxo-4H-pyrrolo[3,2,1-ij]quinoline-5-carboxaldehyde

  • MDL number MFCD01567793
  • Cas Number : 386715-47-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 108-110
Iupac Name 4-oxo-1,2-dihydro-4H-pyrrolo[3,2,1-ij]quinoline-5-carbaldehyde
Inchi Code 1S/C12H9NO2/c14-7-10-6-9-3-1-2-8-4-5-13(11(8)9)12(10)15/h1-3,6-7H,4-5H2
Inchi Key BHQKLXHVZPNJJF-UHFFFAOYSA-N

Description

Synonym 1,2-Dihydro-5-formyl-4-oxo-4H-pyrrolo[3,2,1-ij]quinoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 108-110
Product Link https://store.apolloscientific.co.uk/product/12-dihydro-4-oxo-4h-pyrrolo321-ijquinoline-5-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/12-dihydro-4-oxo-4h-pyrrolo321-ijquinoline-5-carboxaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart