APO455831932 Apollo Scientific Ltd

Ethyl 3-oxo-3-(pyridin-4-yl)propanoate

  • MDL number MFCD00094021
  • Cas Number : 26377-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-59
Synonym Ethyl isonicotinoylacetate, Ethyl 3-oxo-3-(pyridin-4-yl)propionate
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-3-oxo-3-pyridin-4-ylpropanoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-3-oxo-3-pyridin-4-ylpropanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart