APO455831933 Apollo Scientific Ltd

3-(Dimethylamino)piperidine

  • MDL number MFCD06796405
  • Cas Number : 50534-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethylpiperidin-3-amine
Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 58/6.75mm
Product Link https://store.apolloscientific.co.uk/product/3-dimethylaminopiperidine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-dimethylaminopiperidine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart