APO455831948 Apollo Scientific Ltd

5-Bromo-4-chloro-2-methoxybenzoic acid

  • MDL number MFCD09878363
  • Cas Number : 95383-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-carboxy-5-chloroanisole, 5-Bromo-4-chloro-o-anisic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42564

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42564
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart