APO455831952 Apollo Scientific Ltd

1-[(4-Methylphenyl)sulphonyl]-1H-imidazole-4-carboxylic acid

  • MDL number MFCD09878406
  • Cas Number : 957063-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Tosyl-1H-imidazole-4-carboxylic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43234

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43234
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart