APO455831953 Apollo Scientific Ltd

8-Bromo-6-methyl[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD09878402
  • Cas Number : 957062-94-1
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-6-methyl[1,2,4]triazolo[1,5-a]pyridine
Inchi Code 1S/C7H6BrN3/c1-5-2-6(8)7-9-4-10-11(7)3-5/h2-4H,1H3
Inchi Key RVTIPKPMNMOVHB-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-6-methyl124triazolo15-apyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-6-methyl124triazolo15-apyridine
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart