APO455831955 Apollo Scientific Ltd

1-Acetyl-6-chloro-1H-indazole

  • MDL number MFCD09878373
  • Cas Number : 708-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym 1-(6-Chloro-1H-indazol-1-yl)ethan-1-one, 2-(6-Chloro-1H-indazol-1-yl)-2-oxoethane
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43256

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43256
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart