APO455831963 Apollo Scientific Ltd

Imidazo[1,2-a]pyridine-7-carboxylic acid

  • MDL number MFCD09834781
  • Cas Number : 648423-85-2
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 244-247
Iupac Name imidazo[1,2-a]pyridine-7-carboxylic acid
Inchi Code 1S/C8H6N2O2/c11-8(12)6-1-3-10-4-2-9-7(10)5-6/h1-5H,(H,11,12)
Inchi Key TUKDVVKJZVLVTD-UHFFFAOYSA-N

Description

Synonym 7-Carboxyimidazo[1,2-a]pyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 244-247
Product Link https://store.apolloscientific.co.uk/product/imidazo12-apyridine-7-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/imidazo12-apyridine-7-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart