APO455831971 Apollo Scientific Ltd

2,3-Dichlorophenylhydrazine hydrochloride

  • MDL number MFCD00052265
  • Cas Number : 21938-47-6
  • Molecular Weight : 213.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp ca 204(dec.)
Iupac Name 1-(2,3-dichlorophenyl)hydrazine hydrochloride
Inchi Code 1S/C6H6Cl2N2.ClH/c7-4-2-1-3-5(10-9)6(4)8;/h1-3,10H,9H2;1H
Inchi Key RHNATWVYOKUFLK-UHFFFAOYSA-N

Description

Synonym 1,2-Dichloro-3-hydrazinobenzene hydrochloride
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point ca 204(dec.)
Product Link https://store.apolloscientific.co.uk/product/23-dichlorophenylhydrazine-hydrochloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/23-dichlorophenylhydrazine-hydrochloride
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart