APO455831976 Apollo Scientific Ltd

4-Nitro-1H-pyrazole

  • MDL number MFCD00159626
  • Cas Number : 2075-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-164
Purity 98
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 323
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43829

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43829
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart