APO455831981 Apollo Scientific Ltd

4-Chloro-3-methylphenyl isocyanate

  • MDL number MFCD11101408
  • Cas Number : 51488-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-5-(isocyanato)toluene, 1-Chloro-4-isocyanato-2-methylbenzene
Purity 98
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43843

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43843
pictograms GHS06;GHS08
hstatements H319;H331;H335;EUH204
Availability details for
Quantity
  • Availability
  • Add to Cart