APO455831983 Apollo Scientific Ltd

Ethyl 4-(methylsulphanyl)phenylacetate

  • MDL number MFCD09751249
  • Cas Number : 14062-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-58
Synonym 4-(Methylthio)phenylacetic acid ethyl ester, Ethyl [4-(methylsulphanyl)phenyl]acetate, Ethyl 2-[4-(methylsulphanyl)phenyl]ethanoate
Purity 99
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43865

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43865
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart