APO455831984 Apollo Scientific Ltd

5-Chloropyrazine-2-carboxylic acid

  • MDL number MFCD09033269
  • Cas Number : 36070-80-1
  • Molecular Weight : 158.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-160
Synonym 2-Carboxy-5-chloropyrazine, 2-Carboxy-5-chloro-1,4-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 330
Product Link https://store.apolloscientific.co.uk/product/5-chloropyrazine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloropyrazine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart