APO455831987 Apollo Scientific Ltd

6-Chloropyridine-2-boronic acid, pinacol ester

  • MDL number MFCD06798259
  • Cas Number : 652148-92-0
  • Molecular Weight : 239.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 109-111
Iupac Name 2-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C11H15BClNO2/c1-10(2)11(3,4)16-12(15-10)8-6-5-7-9(13)14-8/h5-7H,1-4H3
Inchi Key CROJXBLQCOIOBI-UHFFFAOYSA-N

Description

Synonym 6-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 109-111
Product Link https://store.apolloscientific.co.uk/product/6-chloropyridine-2-boronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloropyridine-2-boronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart