APO455831989 Apollo Scientific Ltd

Uracil-5-carbonitrile

  • MDL number MFCD00051958
  • Cas Number : 4425-56-3
  • Molecular Weight : 137.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >300
Synonym 5-Cyanouracil, 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile, 5-Cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/uracil-5-carbonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/uracil-5-carbonitrile
pictograms GHS06
hstatements H301;H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart