APO455831990 Apollo Scientific Ltd

2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide

  • MDL number MFCD08272105
  • Cas Number : 443956-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Amino-5-bromopyridin-3-yl)methanol hydrobromide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44154

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44154
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart