APO455831997 Apollo Scientific Ltd

N-[(5-Methyl-1,2-oxazol-3-yl)carbamothioyl]benzamide

  • MDL number MFCD00126665
  • Cas Number : 85156-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Synonym 1-Benzoyl-3-(5-methylisoxazol-3-yl)thiourea
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31204

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=31204
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart