APO455832016 Apollo Scientific Ltd

3-Methylpyrazine-2-carboxylic acid

  • MDL number MFCD08705765
  • Cas Number : 41110-28-5
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-177
Synonym 2-Carboxy-3-methylpyrazine, 2-Carboxy-3-methyl-1,4-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methylpyrazine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylpyrazine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart