APO455832020 Apollo Scientific Ltd

2-(Methylsulphonyl)pyrimidine

  • MDL number MFCD06795939
  • Cas Number : 14161-09-2
  • Molecular Weight : 158.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-75
Synonym Methyl pyrimidin-2-yl sulphone
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methylsulphonylpyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-methylsulphonylpyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart