APO455832027 Apollo Scientific Ltd

5-(Aminomethyl)-1,3-oxazole monohydrochloride

  • MDL number MFCD06804578
  • Cas Number : 1196156-45-2
  • Molecular Weight : 134.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-oxazol-5-ylmethanamine hydrochloride
Inchi Code 1S/C4H6N2O.ClH/c5-1-4-2-6-3-7-4;/h2-3H,1,5H2;1H
Inchi Key GWGCSOXRJFLPEH-UHFFFAOYSA-N

Description

Synonym (1,3-Oxazol-5-yl)methylamine monohydrochloride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-aminomethyl-13-oxazole-monohydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminomethyl-13-oxazole-monohydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart