APO455832031 Apollo Scientific Ltd

2-Chloro-7,8-dihydropteridin-6-(5H)-one

  • MDL number MFCD08062762
  • Cas Number : 944580-73-8
  • Molecular Weight : 184.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-oxo-5,6,7,8-tetrahydropteridine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-78-dihydropteridin-6-5h-one

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-chloro-78-dihydropteridin-6-5h-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart