APO455832032 Apollo Scientific Ltd

2-Bromo-4-chlorothieno[3,2-c]pyridine

  • MDL number MFCD08273916
  • Cas Number : 28948-61-0
  • Molecular Weight : 248.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 119
Iupac Name 2-bromo-4-chlorothieno[3,2-c]pyridine
Inchi Code 1S/C7H3BrClNS/c8-6-3-4-5(11-6)1-2-10-7(4)9/h1-3H
Inchi Key QPGCPWZFCLBFCG-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 119
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-chlorothieno32-cpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-chlorothieno32-cpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart