APO455832033 Apollo Scientific Ltd

Pyridazine-4-carboxaldehyde

  • MDL number MFCD06227447
  • Cas Number : 50901-42-3
  • Molecular Weight : 108.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 90-91
Iupac Name 4-pyridazinecarbaldehyde
Inchi Code 1S/C5H4N2O/c8-4-5-1-2-6-7-3-5/h1-4H
Inchi Key BZORCBQGFKOHMY-UHFFFAOYSA-N

Description

Synonym 4-Formylpyridazine, 4-Formyl-1,2-diazine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 90-91
Product Link https://store.apolloscientific.co.uk/product/pyridazine-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/pyridazine-4-carboxaldehyde
pictograms GHS07
hstatements H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart