APO455832037 Apollo Scientific Ltd

6-(Hydroxymethyl)pyridine-2-carboxaldehyde

  • MDL number MFCD10700214
  • Cas Number : 39621-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-6-(hydroxymethyl)pyridine, (6-Formylpyridin-2-yl)methanol
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44178

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44178
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart