APO455832042 Apollo Scientific Ltd

Cyclopropylacetic acid

  • MDL number MFCD00041544
  • Cas Number : 5239-82-7
  • Molecular Weight : 100.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 93
Iupac Name cyclopropylacetic acid
density 1.076
Inchi Code 1S/C5H8O2/c6-5(7)3-4-1-2-4/h4H,1-3H2,(H,6,7)
Inchi Key KVVDRQDTODKIJD-UHFFFAOYSA-N

Description

Melting Point 93
Synonym (Carboxymethyl)cyclopropane, 2-Cyclopropylethanoic acid
Density 1.076
Flash Point 189-191
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/cyclopropylacetic-acid

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/cyclopropylacetic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart