APO455832045 Apollo Scientific Ltd

4-(Dimethylcarbamoyl)benzeneboronic acid, pinacol ester

  • MDL number MFCD05863908
  • Cas Number : 400727-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44234

Safety Information

signalword Danger
pcodes P280;P304+P340;P305+P351+P338
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44234
pictograms GHS07;GHS08
hstatements EUH208;H315;H319;H334;H335
Availability details for
Quantity
  • Availability
  • Add to Cart