APO455832049 Apollo Scientific Ltd

2-Bromo-5-methoxyaniline

  • MDL number MFCD00070741
  • Cas Number : 59557-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Amino-4-bromoanisole, 6-Bromo-m-anisidine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-methoxyaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-methoxyaniline
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart