APO455832051 Apollo Scientific Ltd

Methyl pyrimidine-2-carboxylate

  • MDL number MFCD03426929
  • Cas Number : 34253-03-7
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 101-106
Iupac Name methyl 2-pyrimidinecarboxylate
Inchi Code 1S/C6H6N2O2/c1-10-6(9)5-7-3-2-4-8-5/h2-4H,1H3
Inchi Key JOQJEWAXHQDQAG-UHFFFAOYSA-N

Description

Synonym 2-(Methoxycarbonyl)pyrimidine, 2-(Methoxycarbonyl)-1,3-diazine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 101-106
Product Link https://store.apolloscientific.co.uk/product/methyl-pyrimidine-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-pyrimidine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart