APO455832055 Apollo Scientific Ltd

5-(Fur-2-yl)pyridine-2-carboxylic acid

  • MDL number MFCD10700052
  • Cas Number : 930110-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-165
Synonym 5-(Fur-2-yl)picolinic acid, 2-Carboxy-5-(fur-2-yl)pyridine
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44269

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44269
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart