APO455832058 Apollo Scientific Ltd

7-Amino-1-methyl-1H-indazole

  • MDL number MFCD09870043
  • Cas Number : 41926-06-1
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 131-133
Iupac Name 1-methyl-1H-indazol-7-ylamine
Inchi Code 1S/C8H9N3/c1-11-8-6(5-10-11)3-2-4-7(8)9/h2-5H,9H2,1H3
Inchi Key FKQDTYKVWIDGNT-UHFFFAOYSA-N

Description

Synonym 1-Methyl-1H-indazol-7-amine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 131-133
Product Link https://store.apolloscientific.co.uk/product/7-amino-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-amino-1-methyl-1h-indazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart