APO455832061 Apollo Scientific Ltd

4-(5-Methyl-1,3,4-oxadiazol-2-yl)benzaldehyde

  • MDL number MFCD11109323
  • Cas Number : 179056-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Synonym 2-(4-Formylphenyl)-5-methyl-1,3,4-oxadiazole
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=44323

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=44323
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart