APO455832062 Apollo Scientific Ltd

(Morpholin-4-yl)benzonitrile

  • MDL number MFCD00100115
  • Cas Number : 10282-31-2
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 85
Iupac Name 4-(4-morpholinyl)benzonitrile
Inchi Code 1S/C11H12N2O/c12-9-10-1-3-11(4-2-10)13-5-7-14-8-6-13/h1-4H,5-8H2
Inchi Key ZSCUWVQXQDCSRV-UHFFFAOYSA-N

Description

Synonym 4-(4-Cyanobenzene)morpholine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 85
Product Link https://store.apolloscientific.co.uk/product/morpholin-4-ylbenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/morpholin-4-ylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart