APO455832082 Apollo Scientific Ltd

2-Amino-4,6-dichloropyridine

  • MDL number MFCD04037218
  • Cas Number : 116632-24-7
  • Molecular Weight : 163.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 108-113
Iupac Name 4,6-dichloro-2-pyridinylamine
Inchi Code 1S/C5H4Cl2N2/c6-3-1-4(7)9-5(8)2-3/h1-2H,(H2,8,9)
Inchi Key AGJMDETXSYICGZ-UHFFFAOYSA-N

Description

Synonym 4,6-Dichloropyridin-2-amine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 108-113
Product Link https://store.apolloscientific.co.uk/product/2-amino-46-dichloropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-46-dichloropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart